Copper ≥99.9985% (metals basis), wire, Puratronic®, hard, Ø 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals

10971-G6 10971-H2
CAAA10971-G6EA 184.86 CAD
CAAA10971-G6 CAAA10971-H2
Copper ≥99.9985% (metals basis), wire, Puratronic®, hard, Ø 0.1 mm (0.004 in)
Copper

MDL: MFCD00010965


Purity: 99.9985% CAS No.: 7440-50-8 Physical Form: ≈0.07g/m Appearance: Copper Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR