Palladium ≥99.99%, wire, Premion®, Ø 0.25 mm

Supplier: Thermo Scientific Chemicals

10962-G1 10962-G5 10962-G9
CAAA10962-G1EA 314.42 CAD
CAAA10962-G1 CAAA10962-G5 CAAA10962-G9
Palladium ≥99.99%, wire, Premion®, Ø 0.25 mm
Palladium

MDL: MFCD00011167


Purity: 99.99% CAS No.: 7440-05-3 Physical Form: ≈0.6g/m Odor: Odorless MDL No.: MFCD00011167

Formula: Pd
MW: 106.420 g/mol
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR