Platinum ≥99.997% (metals basis), wire, Premion®, Ø 0.1 mm (0.004 in)
Supplier: Thermo Scientific Chemicals
10959-G1
10959-G5
CAAA10959-G1EA
206.98
CAD
CAAA10959-G1
CAAA10959-G5
Platinum ≥99.997% (metals basis), wire, Premion®, Ø 0.1 mm (0.004 in)
Platinum
MDL: MFCD00011179
Purity: 99.997%
CAS No.: 7440-06-4
Physical Form: ≈0.17g/m
Odor: Odorless
MDL No.: MFCD00011179
Formula:
Pt Boiling Pt: 3827 °C (1013 hPa) Melting Pt: 1769 °C Density: 21.447 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 EINECS: 231-116-1 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...