Platinum ≥99.997% (metals basis), wire, Premion®, Ø 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

10957-BS 10957-BW 10957-CB
CAAA10957-BSEA 311.26 CAD
CAAA10957-BS CAAA10957-BW CAAA10957-CB
Platinum ≥99.997% (metals basis), wire, Premion®, Ø 0.5 mm (0.02 in)
Platinum

MDL: MFCD00011179


Purity: 99.997% CAS No.: 7440-06-4 Physical Form: ≈4.21g/m Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR