Copper ≥99.9999% (metals basis), shot, Puratronic® ≤ 6 mm (0.2 in)

Supplier: Thermo Scientific Chemicals

10955-14 10955-22 10955-36
CAAA10955-14EA 192.76 CAD
CAAA10955-14 CAAA10955-22 CAAA10955-36
Copper ≥99.9999% (metals basis), shot, Puratronic® ≤ 6 mm (0.2 in)
Copper

MDL: MFCD00010965


Purity: 99.9999% CAS No.: 7440-50-8 Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR