Copper ≥99.999% (metals basis), shot, Puratronic® 4-6 mm (0.16-0.24 in)

Supplier: Thermo Scientific Chemicals

10953-18 10953-30 10953-A1
CAAA10953-18EA 255.96 CAD
CAAA10953-18 CAAA10953-30 CAAA10953-A1
Copper ≥99.999% (metals basis), shot, Puratronic® 4-6 mm (0.16-0.24 in)
Copper

MDL: MFCD00010965


Purity: 99.999% CAS No.: 7440-50-8 Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR