Copper ≥99.9995% (metals basis), discs, Ø 9.0+/-0.127 mm (0.354+/-0.005 in), Thickness: 0.8+/-0.051 mm (0.03+/-0.002 in)
Supplier: Thermo Scientific Chemicals
10951-09
10951-18
CAAA10951-09EA
281.24
CAD
CAAA10951-09
CAAA10951-18
Copper ≥99.9995% (metals basis), discs, Ø 9.0+/-0.127 mm (0.354+/-0.005 in), Thickness: 0.8+/-0.051 mm (0.03+/-0.002 in)
Copper
MDL: MFCD00010965
Purity: 99.9995%
CAS No.: 7440-50-8
Physical Form: ≈0.5g/disc
Odor: Odorless
MDL No.: MFCD00010965
Formula:
Cu Boiling Pt: 2595 °C (1013 hPa) Melting Pt: 1083 °C Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...