Cobalt ≥99.995% (metals basis), wire, Puratronic®, Ø 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
10948-G1
10948-G5
CAAA10948-G1EA
262.28
CAD
CAAA10948-G1
CAAA10948-G5
Cobalt ≥99.995% (metals basis), wire, Puratronic®, Ø 0.25 mm (0.01 in)
Cobalt
MDL: MFCD00010935
Purity: 99.995%
CAS No.: 7440-48-4
Physical Form: ≈0.44g/m
Odor: Odorless
MDL No.: MFCD00010935
Formula:
Co Boiling Pt: 2870 °C (1013 hPa) Melting Pt: 1495 °C Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...