Iron ≥99.995% (metals basis), wire, Puratronic®, Ø 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
10937-G1
10937-G5
10937-G9
CAAA10937-G1EA
151.52
CAD
CAAA10937-G1
CAAA10937-G5
CAAA10937-G9
Iron ≥99.995% (metals basis), wire, Puratronic®, Ø 0.25 mm (0.01 in)
Iron
MDL: MFCD00010999
Purity: 99.995%
CAS No.: 7439-89-6
Physical Form: ≈0.39g/m
MDL No.: MFCD00010999
Formula:
Fe Boiling Pt: 3000 °C (1013 hPa) Melting Pt: 1535 °C Density: 7.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010999 CAS Number: 7439-89-6 EINECS: 231-096-4 Merck Index: 13,05109 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...