Charcoal activated ≤ 2 mm
Supplier: Thermo Scientific Chemicals
10926-22
10926-30
10926-A1
CAAA10926-22EA
57.51
CAD
CAAA10926-22
CAAA10926-30
CAAA10926-A1
Charcoal activated ≤ 2 mm
Charcoal activated
For liquid purification
MDL No.: MFCD00133992
For liquid purification
Formula:
C Melting Pt: 3550 °C Density: 2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133992 CAS Number: 7440-44-0 EINECS: 231-153-3 UN: 1362 ADR: 4.2,III REACH: 01-2119488894-16 Merck Index: 13,01818 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...