Boron carbide 90+%

Supplier: Thermo Scientific Chemicals
Warning

10922-22 10922-36
CAAA10922-22EA 90.38 CAD
CAAA10922-22 CAAA10922-36
Boron carbide 90+%
Boron carbide

MDL: MFCD00011520


Purity: 90+% CAS No.: 12069-32-8 Molecular Formula: B4C Formula Weight: 55.26 Physical Form: 22-59 Micron Powder Density: 2.52 Appearance: Black Odor: Odorless Melting Point: 2350° Merck Reference: 14,1344 MDL No.: MFCD00011520

Formula: CB₄
MW: 55.25
Boiling Pt: ∼3500 °C (1013 hPa)
Melting Pt: 2350
Density: 2.52
Storage Temperature: Ambient
MDL Number: MFCD00011520
CAS Number: 12069-32-8
EINECS: 235-111-5
Merck Index: 14,1344

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR