Ytterbium(III) oxide ≥99.998% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

10919-06 10919-14
CAAA10919-06EA 128.93 CAD
CAAA10919-06 CAAA10919-14
Ytterbium(III) oxide ≥99.998% (REO, rare earth oxide basis), REacton®
Ytterbium (III) oxide

MDL: MFCD00011290


Purity: 99.998% CAS No.: 1314-37-0 Molecular Formula: Yb2O3 Formula Weight: 394.08 Physical Form: Powder Density: 9.2 Melting Point: 2227° Merck Reference: 14,10105 MDL No.: MFCD00011290

Formula: O₃Yb₂
Melting Pt: 2227 °C
Density: 9.2
Storage Temperature: Ambient
MDL Number: MFCD00011290
CAS Number: 1314-37-0
EINECS: 215-234-0
Merck Index: 14,10105

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR