Thulium(III) oxide ≥99.997% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

10918-03 10918-06
CAAA10918-03EA 173.8 CAD
CAAA10918-03 CAAA10918-06
Thulium(III) oxide ≥99.997% (REO, rare earth oxide basis), REacton®
Thulium (III) oxide

MDL: MFCD00011285


Purity: 99.997% CAS No.: 12036-44-1 Molecular Formula: O3Tm2 Formula Weight: 385.87 Physical Form: Powder Appearance: Pale green Melting Point: 2425° Merck Reference: 14,9394 MDL No.: MFCD00011285

Formula: O₃Tm₂
MW: 385.87 g/mol
Melting Pt: 2410
Density: 8.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011285
CAS Number: 12036-44-1
EINECS: 234-851-6
Merck Index: 14,9394

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR