Discandium trioxide ≥99.998% (metals basis), REacton® (REM ≤ 50 ppm)
Supplier: Thermo Scientific Chemicals
10916-03
10916-06
CAAA10916-03EA
176.96
CAD
CAAA10916-03
CAAA10916-06
Discandium trioxide ≥99.998% (metals basis), REacton® (REM ≤ 50 ppm)
Discandium trioxide
MDL: MFCD00011223
Purity: 99.998%
CAS No.: 12060-08-1
Molecular Formula: Sc2O3
Formula Weight: 137.91
Physical Form: Powder
Refractive Index: 1.964
Odor: Odorless
Melting Point: 2485°
Merck Reference: 14,8392
MDL No.: MFCD00011223
Formula:
O₃Sc₂ Melting Pt: 2485 °C Density: 3.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011223 CAS Number: 12060-08-1 EINECS: 235-042-0 Merck Index: 14,8392 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...