Samarium(III) oxide ≥99.998% (metals basis), REacton® (REM ≤ 100 ppm)
Supplier: Thermo Scientific Chemicals
10915-06
10915-14
10915-22
CAAA10915-06EA
130.82
CAD
CAAA10915-06
CAAA10915-14
CAAA10915-22
Samarium(III) oxide ≥99.998% (metals basis), REacton® (REM ≤ 100 ppm)
Samarium (III) oxide
MDL: MFCD00011237
Purity: 99.998%
CAS No.: 12060-58-1
Molecular Formula: Sm2O3
Formula Weight: 348.70
Physical Form: Powder
Refractive Index: 1.97
Melting Point: 2300°
Merck Reference: 14,8347
MDL No.: MFCD00011237
Formula:
O₃Sm₂ Melting Pt: 2300 °C Density: 7.68 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011237 CAS Number: 12060-58-1 EINECS: 235-043-6 Merck Index: 14,8347 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...