Lutetium(III) oxide ≥99.995% (metals basis), REacton® (REM ≤ 50 ppm)
Supplier: Thermo Scientific Chemicals
|
Warning
|
10912-03
10912-06
CAAA10912-03EA
240.16
CAD
CAAA10912-03
CAAA10912-06
Lutetium(III) oxide ≥99.995% (metals basis), REacton® (REM ≤ 50 ppm)
Lutetium (III) oxide
MDL: MFCD00011100
Purity: 99.995%
CAS No.: 12032-20-1
Molecular Formula: Lu2O3
Formula Weight: 397.94
Physical Form: Powder
Density: 9.420
Melting Point: 2487°
Merck Reference: 14,5615
MDL No.: MFCD00011100
Formula:
Lu₂O₃ Melting Pt: 2487 °C Density: 9.420 Storage Temperature: Ambient |
MDL Number:
MFCD00011100 CAS Number: 12032-20-1 EINECS: 234-764-3 Merck Index: 14,5615 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...