Holmium(III) oxide ≥99.995% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

10910-03 10910-06 10910-14
CAAA10910-03EA 88.01 CAD
CAAA10910-03 CAAA10910-06 CAAA10910-14
Holmium(III) oxide ≥99.995% (REO, rare earth oxide basis), REacton®
Holmium (III) oxide

MDL: MFCD00011053


Purity: 99.995% CAS No.: 12055-62-8 Molecular Formula: Ho2O3 Formula Weight: 377.86 Physical Form: Powder Appearance: Pale brown Odor: Odorless Melting Point: 2360° Merck Reference: 14,4727 MDL No.: MFCD00011053

Formula: Ho₂O₃
Boiling Pt: 2600 °C (1013 hPa)
Melting Pt: 2360 °C
Density: 3.79 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011053
CAS Number: 12055-62-8
EINECS: 235-015-3
Merck Index: 14,4727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR