Titanium(IV) oxide ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
10897-09
10897-18
10897-30
CAAA10897-09EA
366.56
CAD
CAAA10897-09
CAAA10897-18
CAAA10897-30
Titanium(IV) oxide ≥99.995% (metals basis), Puratronic®
Titanium (IV) oxide
MDL: MFCD00011269
Purity: 99.995%
CAS No.: 13463-67-7
Molecular Formula: TiO2, predominantly rutile and anatase
Formula Weight: 79.90
Physical Form: -22 Mesh Powder
Density: 4.26
Appearance: White
Odor: Odorless
Melting Point: 1830-1850°
Boiling Point: 2500-3000°
Merck Reference: 14,9472
MDL No.: MFCD00011269
Formula:
TiO₂ Boiling Pt: 2500…3000 °C Melting Pt: 1830…1850 °C Density: 4.26 Storage Temperature: Ambient |
MDL Number:
MFCD00011269 CAS Number: 13463-67-7 EINECS: 236-675-5 Merck Index: 14,9472 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...