Tantalum pentachloride ≥99.99% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10880-06
10880-14
10880-22
CAAA10880-06EA
89.43
CAD
CAAA10880-06
CAAA10880-14
CAAA10880-22
Tantalum pentachloride ≥99.99% (metals basis), Puratronic®
Tantalum pentachloride
MDL: MFCD00011253
Fieser: 16,312 17,321 18,343 19,318 20,355 21,407
Purity: 99.99%
CAS No.: 7721-01-9
Molecular Formula: TaCl5
Formula Weight: 358.21
Physical Form: Resublimed Crystalline, Ampouled under argon
Density: 3.68
Appearance: White to pale yellow
Odor: Acidic
Melting Point: 216°
Boiling Point: 242°
Merck Reference: 14,9054
UN No.: 3260
MDL No.: MFCD00011253
Formula:
Cl₅Ta Boiling Pt: 242 °C Melting Pt: 216 °C Density: 3.68 Storage Temperature: Ambient |
MDL Number:
MFCD00011253 CAS Number: 7721-01-9 EINECS: 231-755-6 UN: 3260 ADR: 8,III Merck Index: 14,9054 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...