Strontium nitrate ≥99.9965% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10879-14
10879-22
CAAA10879-14EA
214.88
CAD
CAAA10879-14
CAAA10879-22
Strontium nitrate ≥99.9965% (metals basis), Puratronic®
Strontium nitrate
MDL: MFCD00011248
Purity: 99.9965%
CAS No.: 10042-76-9
Molecular Formula: N2O6Sr
Formula Weight: 211.63
Physical Form: Powder, Packaged under argon
Density: 2.990
Appearance: White
Odor: Odorless
Melting Point: 570°
Merck Reference: 14,8845
UN No.: 1507
MDL No.: MFCD00011248
Formula:
Sr(NO₃)₂ Boiling Pt: 645 °C (1013 hPa) Melting Pt: 570 °C Density: 2.990 Storage Temperature: Ambient |
MDL Number:
MFCD00011248 CAS Number: 10042-76-9 EINECS: 233-131-9 UN: 1507 ADR: 5.1,III Merck Index: 14,8845 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...