Strontium fluoride ≥99.99% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

10878-06 10878-14 10878-22
CAAA10878-06EA 92.9 CAD
CAAA10878-06 CAAA10878-14 CAAA10878-22
Strontium fluoride ≥99.99% (metals basis), Puratronic®
Strontium fluoride

MDL: MFCD00011251


Purity: 99.99% CAS No.: 7783-48-4 Molecular Formula: SrF2 Formula Weight: 125.62 Physical Form: Powder Density: 4.240 Refractive Index: 1.442 Appearance: White Odor: Odorless Melting Point: 1473° Merck Reference: 14,8842 MDL No.: MFCD00011251

Formula: F₂Sr
Boiling Pt: 2460 °C (1013 hPa)
Melting Pt: 1473 °C
Density: 4.240
Storage Temperature: Ambient
MDL Number: MFCD00011251
CAS Number: 7783-48-4
EINECS: 232-000-3
UN: 3288
ADR: 6.1,III
Merck Index: 14,8842

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR