di-Sodium tetraborate, anhydrous ≥99.998% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Borax, Sodium tetraborate

10875-14 10875-22 10875-36
CAAA10875-14EA 139.04 CAD
CAAA10875-14 CAAA10875-22 CAAA10875-36
di-Sodium tetraborate, anhydrous ≥99.998% (metals basis), Puratronic®
di-Sodium tetraborate

MDL: MFCD00081185


Purity: 99.998% CAS No.: 1330-43-4 Molecular Formula: B4Na2O7 Formula Weight: 201.22 Physical Form: Powder Density: 2.367 Refractive Index: 1.501 Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 741° Boiling Point: 1575° dec. Merck Reference: 14,8590 MDL No.: MFCD00081185

Formula: Na₂B₄O₇
MW: 201.22 g/mol
Boiling Pt: 1575 (dec.)
Melting Pt: 742
Density: 2.367
Storage Temperature: Ambient
MDL Number: MFCD00081185
CAS Number: 1330-43-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 14,8590

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR