Sodium sulfate ≥99.9955% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
Synonyms:
Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt
10874-14
10874-22
CAAA10874-14EA
360.24
CAD
CAAA10874-14
CAAA10874-22
Sodium sulfate ≥99.9955% (metals basis), Puratronic®
Sodium sulphate
MDL: MFCD00003504
Fieser: 1,1103
Purity: 99.9955%
CAS No.: 7757-82-6
Molecular Formula: Na2SO4
Formula Weight: 142.04
Physical Form: Powder
Density: 2.680
Refractive Index: 1.484
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 884°
Merck Reference: 14,8680
MDL No.: MFCD00003504
Formula:
Na₂SO₄ Boiling Pt: 1429 °C (1013 hPa) Melting Pt: 884 °C Density: 2.680 Storage Temperature: Ambient |
MDL Number:
MFCD00003504 CAS Number: 7757-82-6 EINECS: 231-820-9 Merck Index: 14,8680 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...