Sodium iodide dihydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium iodide hydrate
10871-06
10871-14
10871-22
CAAA10871-06EA
82.48
CAD
CAAA10871-06
CAAA10871-14
CAAA10871-22
Sodium iodide dihydrate ≥99.999% (metals basis), Puratronic®
Sodium iodide dihydrate
MDL: MFCD00003532
Purity: 99.999%
Molecular Formula: NaI.xH2O
Formula Weight: 149.89(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Formula:
INa·2H₂O MW: 185.92 Melting Pt: 64.3 Density: 2.67 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149211 CAS Number: 13517-06-1 EINECS: 231-679-3 UN: 3077 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...