Sodium dihydrogen phosphate monohydrate ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate
MDL: MFCD00149208
Fieser: 1,395
Purity: 99.998%
CAS No.: 10049-21-5
Molecular Formula: H2NaO4P.H2O
Formula Weight: 137.99 (119.98anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 100° -H2O
Merck Reference: 14,8660
MDL No.: MFCD00149208
Formula:
NaH₂PO₄·H₂O MW: 137.99 g/mol Melting Pt: 100 °C (-H₂O) Density: 2.04 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149208 CAS Number: 10049-21-5 EINECS: 231-449-2 Merck Index: 14,8660 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...