Sodium hydrogen carbonate ≥99.998% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium bicarbonate

10863-09 10863-18
CAAA10863-09EA 165.9 CAD
CAAA10863-09 CAAA10863-18
Sodium hydrogen carbonate ≥99.998% (metals basis), Puratronic®
Sodium hydrogen carbonate

MDL: MFCD00003528 Beilstein Registry No.: 4153970 Notes: m.p. about 50°C, begins to lose CO2. about 100°C, converts to Na2CO3 Fieser: 3,260 5,595


Purity: 99.998% CAS No.: 144-55-8 Molecular Formula: CHNaO3 Formula Weight: 84.01 Physical Form: Powder Density: 2.159 Refractive Index: 1.500 Appearance: White Odor: Odorless pH: 8,1-8,4 Beilstein Reference: 4153970 Merck Reference: 14,8583 MDL No.: MFCD00003528

Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270
Density: 2.159
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 14,8583
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR