Silver chloride ≥99.997% (metals basis), Premion®

Supplier: Thermo Scientific Chemicals
Warning

10857-06 10857-14 10857-22
CAAA10857-06EA 110.76 CAD
CAAA10857-06 CAAA10857-14 CAAA10857-22
Silver chloride ≥99.997% (metals basis), Premion®
Silver chloride

MDL: MFCD00003399


Purity: 99.997% CAS No.: 7783-90-6 Molecular Formula: AgCl Formula Weight: 143.32 Physical Form: Coarse Powder Density: 5.56 Refractive Index: 2.071 Sensitivity: Light Sensitive Appearance: White Odor: Odorless Melting Point: 455° Boiling Point: 1550° Merck Reference: 14,8509 UN No.: 3077 MDL No.: MFCD00003399

Formula: AgCl
MW: 143.32 g/mol
Boiling Pt: 1550 °C
Melting Pt: 455 °C
Density: 5.56
Storage Temperature: Ambient
MDL Number: MFCD00003399
CAS Number: 7783-90-6
EINECS: 232-033-3
UN: 3077
ADR: 9,III
Merck Index: 14,8509

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR