Selenious acid ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10855-14
10855-22
CAAA10855-14EA
510.34
CAD
CAAA10855-14
CAAA10855-22
Selenious acid ≥99.999% (metals basis), Puratronic®
Selenious acid
MDL: MFCD00011331
Purity: 99.999%
CAS No.: 7783-00-8
Molecular Formula: H2SeO3
Formula Weight: 128.97
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Merck Reference: 14,8430
UN No.: 3290
MDL No.: MFCD00011331
Formula:
H₂O₃Se Boiling Pt: 315 °C (1013 hPa) Melting Pt: 70 °C (dec.) Density: 1.998 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011331 CAS Number: 7783-00-8 EINECS: 231-974-7 UN: 3290 ADR: 6.1,II Merck Index: 14,8430 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...