Selenium dioxide ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

10854-06 10854-14
CAAA10854-06EA 127.98 CAD
CAAA10854-06 CAAA10854-14
Selenium dioxide ≥99.999% (metals basis), Puratronic®
Selenium dioxide

MDL: MFCD00003562


Purity: 99.999% CAS No.: 7446-08-4 Molecular Formula: SeO2 Formula Weight: 110.96 Physical Form: Powder/Lump Odor: Odorless Merck Reference: 14,8434 UN No.: 3283 MDL No.: MFCD00003562

Formula: SeO₂
MW: 110.96 g/mol
Melting Pt: 315 (subl.)
Density: 3.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003562
CAS Number: 7446-08-4
EINECS: 231-194-7
UN: 3283
ADR: 6.1 II
Merck Index: 14,8434

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR