Rubidium chloride 99.975% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Rubidiumchloride
10851-09
10851-18
CAAA10851-09EA
380.78
CAD
CAAA10851-09
CAAA10851-18
Rubidium chloride 99.975% (metals basis), Puratronic®
Rubidium chloride
MDL: MFCD00011187
Purity: 99.975%
CAS No.: 7791-11-9
Molecular Formula: RbCl
Formula Weight: 120.92
Physical Form: Crystalline Aggregates
Density: 2.800
Refractive Index: 1.493
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 718°
Boiling Point: 1390°
Merck Reference: 14,8284
MDL No.: MFCD00011187
Formula:
RbCl Boiling Pt: 1390 °C Melting Pt: 718 °C Density: 2.800 Storage Temperature: Ambient |
MDL Number:
MFCD00011187 CAS Number: 7791-11-9 EINECS: 232-240-9 Merck Index: 14,8284 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...