Ammonium perrhenate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10846-04
10846-09
CAAA10846-04EA
224.36
CAD
CAAA10846-04
CAAA10846-09
Ammonium perrhenate ≥99.999% (metals basis), Puratronic®
Ammonium perrhenate
MDL: MFCD00012377
Purity: 99.999%
CAS No.: 13598-65-7
Molecular Formula: NH4ReO4
Formula Weight: 268.24
Physical Form: Crystalline
Density: 3.97
Appearance: White
Melting Point: dec.
UN No.: 1479
MDL No.: MFCD00012377
Formula:
H₄NO₄Re Melting Pt: Dec. Density: 3.97 Storage Temperature: Ambient |
MDL Number:
MFCD00012377 CAS Number: 13598-65-7 EINECS: 237-075-6 UN: 1479 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...