Potassium dihydrogen phosphate ≥99.999% (metals basis), Puratronic® (Si ≤ 10 ppm)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

10840-14 10840-22
CAAA10840-14EA 462.94 CAD
CAAA10840-14 CAAA10840-22
Potassium dihydrogen phosphate ≥99.999% (metals basis), Puratronic® (Si ≤ 10 ppm)
Potassium dihydrogen phosphate

MDL: MFCD00011401 Notes: Loses H2O at 400°C, forming metaphosphate


Purity: 99.999% CAS No.: 7778-77-0 Molecular Formula: H2KO4P Formula Weight: 136.09 Physical Form: Crystalline powder Density: 2.338 Appearance: White Melting Point: 253° Merck Reference: 14,7659 MDL No.: MFCD00011401

Formula: KH₂PO₄
MW: 136.09 g/mol
Boiling Pt: 100
Melting Pt: 253
Density: 2.338
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 14,7659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR