Potassium carbonate ≥99.997% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

10838-06 10838-14 10838-22
CAAA10838-06EA 48.98 CAD
CAAA10838-06 CAAA10838-14 CAAA10838-22
Potassium carbonate ≥99.997% (metals basis), Puratronic®
Potassium carbonate

MDL: MFCD00011382 Beilstein Registry No.: 4267587 Fieser: 1,1014 4,50 5,552 8,408 9,382 10,323 11,433 13,254


Purity: 99.997% CAS No.: 584-08-7 Molecular Formula: K2CO3 Formula Weight: 138.21 Physical Form: Powder Density: 2.29 Melting Point: 891° Beilstein Reference: 4267587 Merck Reference: 14,7619 MDL No.: MFCD00011382

Formula: K₂CO₃
Melting Pt: 891 °C
Density: 2.29
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 14,7619
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR