Potassium bromide ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
10837-09
10837-18
CAAA10837-09EA
175.38
CAD
CAAA10837-09
CAAA10837-18
Potassium bromide ≥99.999% (metals basis), Puratronic®
Potassium bromide
MDL: MFCD00011358
Fieser: 21,358
Purity: 99.999%
CAS No.: 7758-02-3
Molecular Formula: BrK
Formula Weight: 119.01
Physical Form: Crystalline
Density: 2.75
Refractive Index: 1.559
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 730°
Boiling Point: 1435°
Merck Reference: 14,7618
MDL No.: MFCD00011358
Formula:
KBr Boiling Pt: 1435 °C Melting Pt: 730 °C Density: 2.75 Storage Temperature: Ambient |
MDL Number:
MFCD00011358 CAS Number: 7758-02-3 EINECS: 231-830-3 Merck Index: 14,7618 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...