Ammonium hexachloroosmate(IV) ≥99.99% (metals basis) Os ≥ 42.8%, Premion®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diammonium hexachloroosmate
10823-03
10823-06
CAAA10823-02EA
340.63
CAD
CAAA10823-02
CAAA10823-03
CAAA10823-06
Ammonium hexachloroosmate(IV) ≥99.99% (metals basis) Os ≥ 42.8%, Premion®
Ammonium hexachloroosmate(IV)
MDL: MFCD00010883
Purity: 99.99%
CAS No.: 12125-08-5
Molecular Formula: H8Cl6N2Os
Formula Weight: 438.99
Physical Form: Powder
Density: 2.9
Appearance: Brown to black
Melting Point: 170° subl.
Merck Reference: 14,536
MDL No.: MFCD00010883
Formula:
H₈Cl₆N₂Os MW: 439.02 g/mol Melting Pt: 170 °C (subl.) Density: 2.9 Storage Temperature: Ambient |
MDL Number:
MFCD00010883 CAS Number: 12125-08-5 EINECS: 235-188-5 Merck Index: 14,536 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...