Nickel(II) nitrate hexahydrate ≥99.9985% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

10816-06 10816-18 10816-30
CAAA10816-06EA 107.12 CAD
CAAA10816-06 CAAA10816-18 CAAA10816-30
Nickel(II) nitrate hexahydrate ≥99.9985% (metals basis), Puratronic®
Nickel (II) nitrate hexahydrate

MDL: MFCD00149805


Purity: 99.9985% CAS No.: 13478-00-7 Molecular Formula: N2NiO6.6H2O Formula Weight: 290.80 (182.70anhy) Physical Form: Crystalline Aggregates Density: 2.05 Appearance: Green Odor: Odorless Melting Point: 56.7° Boiling Point: 137° Merck Reference: 14,6513 UN No.: 2725 MDL No.: MFCD00149805

Formula: Ni(NO₃)₂·6H₂O
MW: 290.79 g/mol
Boiling Pt: 137 °C
Melting Pt: 56.7 °C
Density: 2.05
Storage Temperature: Ambient
MDL Number: MFCD00149805
CAS Number: 13478-00-7
EINECS: 236-068-5
UN: 2725
ADR: 5.1,III
Merck Index: 14,6513

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR