Nickel(II) chloride hydrate ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10815-06
10815-14
10815-22
CAAA10815-06EA
88.01
CAD
CAAA10815-06
CAAA10815-14
CAAA10815-22
Nickel(II) chloride hydrate ≥99.995% (metals basis), Puratronic®
Nickel (II) chloride hydrate
MDL: MFCD00149808
Fieser: 12,336 15,217 21,281
Purity: 99.995%
CAS No.: 69098-15-3
Molecular Formula: Cl2Ni.xH2O
Formula Weight: 129.60(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: Green
Merck Reference: 14,6505
UN No.: 3288
MDL No.: MFCD00149808
Formula:
Cl₂Ni·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149808 CAS Number: 69098-15-3 EINECS: 231-743-0 UN: 3288 ADR: 6.1,III Merck Index: 14,6505 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...