Mercury(II) chloride ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercuric chloride, Mercury Chloride, Mercury dichloride

10808-14 10808-22
CAAA10808-14EA 159.58 CAD
CAAA10808-14 CAAA10808-22
Mercury(II) chloride ≥99.999% (metals basis), Puratronic®
Mercury (II) chloride

MDL: MFCD00011041 Fieser: 1,652 6,359 9,291 12,304 13,175 15,200 18,232 19,211 20,238


Purity: 99.999% CAS No.: 7487-94-7 Molecular Formula: Cl2Hg Formula Weight: 271.50 Physical Form: Powder Density: 5.4 Refractive Index: 1.8590 Appearance: White Odor: Odorless Melting Point: 280-282° Boiling Point: 302° Merck Reference: 14,5876 UN No.: 1624 MDL No.: MFCD00011041

Formula: HgCl₂
MW: 271.5 g/mol
Boiling Pt: 302
Melting Pt: 273
Density: 5.4
Storage Temperature: Ambient
MDL Number: MFCD00011041
CAS Number: 7487-94-7
EINECS: 231-299-8
UN: 1624
ADR: 6.1 II
Merck Index: 14,5876

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR