Manganese(II) nitrate hydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10806-04
10806-09
10806-18
CAAA10806-04EA
40.29
CAD
CAAA10806-04
CAAA10806-09
CAAA10806-18
Manganese(II) nitrate hydrate ≥99.999% (metals basis), Puratronic®
Manganese (II) nitrate hydrate
MDL: MFCD00149788
Purity: 99.999%
CAS No.: 15710-66-4
Molecular Formula: Mn(NO3)2.xH2O
Formula Weight: 178.95(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: Pink
Odor: Odorless
Melting Point: ca 26°
Merck Reference: 14,5733
UN No.: 2724
MDL No.: MFCD00149788
Formula:
Mn(NO₃)₂·H₂O Melting Pt: ∼ 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149788 CAS Number: 15710-66-4 EINECS: 233-828-8 UN: 2724 ADR: 5.1,III Merck Index: 14,5733 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...