Magnesium sulfate hydrate ≥99.997% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals

10801-09 10801-18
CAAA10801-09EA 116.6 CAD
CAAA10801-09 CAAA10801-18
Magnesium sulfate hydrate ≥99.997% (metals basis), Puratronic®
Magnesium sulphate hydrate

MDL: MFCD00149787 Notes: 150°C -6H2O, 200°C -7H2O


Purity: 99.997% CAS No.: 22189-08-8 Molecular Formula: MgSO4.xH2O Formula Weight: 120.37(anhy) Physical Form: Crystalline Aggregates Density: 1.68 Appearance: White Odor: Odorless Merck Reference: 14,5691 MDL No.: MFCD00149787

Formula: MgSO₄·nH₂O
Density: 1.68
Storage Temperature: Ambient
MDL Number: MFCD00149787
CAS Number: 22189-08-8
EINECS: 231-298-2
Merck Index: 14,5691

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR