Magnesium nitrate hydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10799-06
10799-14
10799-22
10799-36
CAAA10799-06EA
49.3
CAD
CAAA10799-06
CAAA10799-14
CAAA10799-22
CAAA10799-36
Magnesium nitrate hydrate ≥99.999% (metals basis), Puratronic®
Magnesium nitrate hydrate
It is used in printing, chemical, agriculture and ceramics industries. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.
Purity: 99.999%
CAS No.: 10377-60-3
Molecular Formula: Mg(NO3)2.xH2O
Formula Weight: 148.32(anhy)
Physical Form: Crystalline Aggregates
Density: 1.46
Melting Point: 89°
Boiling Point: 330° dec.
Merck Reference: 14,5674
UN No.: 1474
MDL No.: MFCD00011103
Formula:
Mg(NO₃)₂·xH₂O Boiling Pt: 330 °C (dec.) Melting Pt: 89 °C Density: 1.46 Flash Pt: 93.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011103 CAS Number: 10377-60-3 EINECS: 233-826-7 UN: 1474 ADR: 5.1,III Merck Index: 14,5674 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...