Magnesium chloride hexahydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
10797-14
10797-22
CAAA10797-14EA
317.58
CAD
CAAA10797-14
CAAA10797-22
Magnesium chloride hexahydrate ≥99.999% (metals basis), Puratronic®
Magnesium chloride hexahydrate
MDL: MFCD00149781
Fieser: 10,251
Purity: 99.999%
CAS No.: 7791-18-6
Molecular Formula: MgCl2.6H2O
Formula Weight: 203.30 (95.22anhy)
Physical Form: Crystalline Aggregates
Density: 1.570
Appearance: White
Odor: Odorless
Melting Point: 117° dec.
Merck Reference: 14,5662
MDL No.: MFCD00149781
Formula:
MgCl₂·6H₂O Boiling Pt: 1412 °C (1013 hPa) Melting Pt: 117 °C (dec.) Density: 1.570 Storage Temperature: Ambient |
MDL Number:
MFCD00149781 CAS Number: 7791-18-6 EINECS: 232-094-6 Merck Index: 14,5662 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...