Magnesium hydroxide carbonate hydrate ≥99.996% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals

Synonyms: Magnesium carbonate basic, Magnesium carbonate hydroxide, Magnesium carbonate hydrated basic

10796-06 10796-14 10796-22
CAAA10796-06EA 116.13 CAD
CAAA10796-06 CAAA10796-14 CAAA10796-22
Magnesium hydroxide carbonate hydrate ≥99.996% (metals basis), Puratronic®
Magnesium hydroxide carbonate hydrate

MDL: MFCD00149783


Purity: 99.996% CAS No.: 39409-82-0 Molecular Formula: MgCO3.Mg(OH)2.xH2O (x≈3) Formula Weight: 142.63(anhy) Physical Form: Powder Appearance: White Melting Point: dec. Merck Reference: 14,5660 MDL No.: MFCD00149783

Formula: MgCO₃·Mg(OH)₂·xH₂O
Boiling Pt: 3600 °C (1013 hPa)
Melting Pt: Dec.
Density: 2.16 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149783
CAS Number: 39409-82-0
EINECS: 235-192-7
Merck Index: 14,5660

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR