Magnesium acetate tetrahydrate ≥99.997% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
10795-14
10795-22
CAAA10795-14EA
199.08
CAD
CAAA10795-14
CAAA10795-22
Magnesium acetate tetrahydrate ≥99.997% (metals basis), Puratronic®
Magnesium acetate tetrahydrate
MDL: MFCD00149214
Beilstein Registry No.: 3730605
Purity: 99.997%
CAS No.: 16674-78-5
Molecular Formula: Mg(CH3COO)2.4H2O
Formula Weight: 214.45 (142.39anhy)
Physical Form: Crystalline
Density: 1.454
Melting Point: 80°
Beilstein Reference: 3730605
Merck Reference: 14,5654
MDL No.: MFCD00149214
Formula:
(H₃CCOO)₂Mg·4H₂O Melting Pt: 80 °C Density: 1.454 Storage Temperature: Ambient |
MDL Number:
MFCD00149214 CAS Number: 16674-78-5 EINECS: 205-554-9 Merck Index: 14,5654 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...