Hafnium ≥99.5% (metals basis excluding Zr), foil (Zr ∼ 3%), annealed, Thickness: 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

10794-FI 10794-GH
CAAA10794-FIEA 699.94 CAD
CAAA10794-FI CAAA10794-GH
Hafnium ≥99.5% (metals basis excluding Zr), foil (Zr ∼ 3%), annealed, Thickness: 1.0 mm (0.04 in)
Hafnium

MDL: MFCD00011032


Purity: 99.5% CAS No.: 7440-58-6 Physical Form: ≈32.8g/50x50mm, Temper: soft Odor: Odorless MDL No.: MFCD00011032

Formula: Hf
MW: 178.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR