Hafnium ≥99.5% (metals basis excluding Zr), foil (Zr ∼ 3%), annealed, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
10793-FJ
10793-FP
CAAA10793-FJEA
541.94
CAD
CAAA10793-FJ
CAAA10793-FP
Hafnium ≥99.5% (metals basis excluding Zr), foil (Zr ∼ 3%), annealed, Thickness: 0.25 mm (0.01 in)
Hafnium
MDL: MFCD00011032
Purity: 99.5%
CAS No.: 7440-58-6
Physical Form: ≈13.3g/50x80mm
Odor: Odorless
MDL No.: MFCD00011032
Formula:
Hf Storage Temperature: Ambient |
MDL Number:
MFCD00011032 CAS Number: 7440-58-6 EINECS: 231-166-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...