Tin 99,95 Metals basis, wire, Ø 3.2 mm (0.13 in)

Supplier: Thermo Scientific Chemicals

10790-G1 10790-G5
CAAA10790-G1EA 157.53 CAD
CAAA10790-G1 CAAA10790-G5
Tin 99,95 Metals basis, wire, Ø 3.2 mm (0.13 in)
Tin

MDL: MFCD00133862


Purity: 99.95% CAS No.: 7440-31-5 Physical Form: ≈58g/m Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 23-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR