Palladium ≥99.995% (metals basis), powder, Premion® , -22 mesh

Supplier: Thermo Scientific Chemicals
Warning

10788-93 10788-03 10788-93 10788-06 10788-03 10788-06
CAAA10788-03EA 522.98 CAD
CAAA10788-03 CAAA10788-93 AA10788-93 AA10788-03 CAAA10788-06 AA10788-06
Palladium ≥99.995% (metals basis), powder, Premion® , -22 mesh
Palladium
Formula: Pd
MW: 106.420 g/mol
Density: 12.02 g/cm³ (20 °C)
MDL Number: MFCD00011167
CAS Number: 7440-05-3
UN: 3089
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR