Aluminium ≥99.999% (metals basis), rods, Puratronic®, Ø 9.5 mm (0.37 in)
Supplier: Thermo Scientific Chemicals
10779-BQ
10779-BU
10779-CZ
CAAA10779-BQEA
113.76
CAD
CAAA10779-BQ
CAAA10779-BU
CAAA10779-CZ
Aluminium ≥99.999% (metals basis), rods, Puratronic®, Ø 9.5 mm (0.37 in)
Aluminium
MDL: MFCD00134029
Purity: 99.999%
CAS No.: 7429-90-5
Physical Form: ≈1.91g/cm
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al Boiling Pt: 2327 °C (1013 hPa) Melting Pt: 660 °C Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...