Lithium ≥99.8%, wire (max. 0.03% Na), Ø 3.2 mm (0.125 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
10771-14
10771-49
CAAA10771-14EA
254.38
CAD
CAAA10771-14
CAAA10771-49
Lithium ≥99.8%, wire (max. 0.03% Na), Ø 3.2 mm (0.125 in)
Lithium
MDL: MFCD00134051
Purity: 99.8%
CAS No.: 7439-93-2
Physical Form: ≈5.8m/25g, Pkg in mylar under argon w/oil coating
Appearance: Silver-grey
Odor: Odorless
UN No.: 1415
MDL No.: MFCD00134051
Formula:
Li MW: 6.94 g/mol Boiling Pt: 1317 Melting Pt: 179 Density: 0.53 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134051 CAS Number: 7439-93-2 EINECS: 231-102-5 UN: 1415 ADR: 4.3 I Merck Index: 13,05542 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...